Assay Details
| Gene Symbol: LOC4347638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755237.2 | XP_015610723.1 | 6 - 7 | 1005 | 70 | |
| GenBank mRNA | AB097460.1 | - | 6 - 7 | 877 | 70 | |
| AK073610.1 | - | 6 - 7 | 949 | 70 | ||
| AK104977.1 | - | 6 - 7 | 893 | 70 | ||
More Information
Set Membership: |
|

LOC4347638
Genomic Map