Assay Details
Gene Symbol: LOC4347787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756522.2 | XP_015612008.1 | 16 - 17 | 2139 | 111 | |
GenBank mRNA | AK066732.1 | - | 16 - 17 | 2146 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
