Assay Details
Gene Symbol: LOC4347787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756522.2 | XP_015612008.1 | 4 - 5 | 819 | 148 | |
GenBank mRNA | AK066732.1 | - | 4 - 5 | 826 | 148 | |
AK073242.1 | - | 4 - 5 | 825 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
