Assay Details
| Gene Symbol: LOC4348496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759277.2 | XP_015614763.1 | 12 - 13 | 1605 | 127 | |
| XM_015759278.2 | XP_015614764.1 | 12 - 13 | 1602 | 127 | ||
| GenBank mRNA | AK066800.1 | - | 12 - 13 | 1603 | 127 | |
| AK099380.1 | - | 12 - 13 | 1504 | 127 | ||
| AK104008.1 | - | 8 - 9 | 911 | 127 | ||
More Information
Set Membership: |
|

LOC4348496
Genomic Map