Assay Details
| Gene Symbol: LOC4348496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759277.2 | XP_015614763.1 | 9 - 10 | 1336 | 102 | |
| XM_015759278.2 | XP_015614764.1 | 9 - 10 | 1336 | 102 | ||
| GenBank mRNA | AK066800.1 | - | 9 - 10 | 1334 | 102 | |
| AK099380.1 | - | 9 - 10 | 1235 | 102 | ||
| AK104008.1 | - | 5 - 6 | 642 | 102 | ||
More Information
Set Membership: |
|

LOC4348496
Genomic Map