Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 11 - 12 | 945 | 87 | |
XM_015758080.2 | XP_015613566.1 | 11 - 12 | 883 | 87 | ||
XM_015758081.2 | XP_015613567.1 | 10 - 11 | 783 | 87 | ||
GenBank mRNA | AB039671.1 | - | 11 - 12 | 923 | 87 | |
AK059912.1 | - | 11 - 12 | 741 | 87 | ||
AK068374.1 | - | 11 - 12 | 938 | 87 | ||
AK068613.1 | - | 11 - 12 | 863 | 87 | ||
BT128921.1 | - | 1 - 2 | 60 | 87 | ||
BT129662.1 | - | 545 | 87 | |||
BT130454.1 | - | 3 - 4 | 215 | 87 | ||
BT132180.1 | - | 1 - 2 | 45 | 87 | ||
BT132273.1 | - | 1 - 2 | 52 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
