Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 12 - 13 | 1032 | 102 | |
XM_015758080.2 | XP_015613566.1 | 12 - 13 | 970 | 102 | ||
XM_015758081.2 | XP_015613567.1 | 11 - 12 | 870 | 102 | ||
GenBank mRNA | AB039671.1 | - | 12 - 13 | 1010 | 102 | |
AK058373.1 | - | 2 - 3 | 105 | 102 | ||
AK059912.1 | - | 12 - 13 | 828 | 102 | ||
AK068374.1 | - | 12 - 13 | 1025 | 102 | ||
AK068613.1 | - | 12 - 13 | 950 | 102 | ||
BT128921.1 | - | 2 - 3 | 147 | 102 | ||
BT130454.1 | - | 4 - 5 | 302 | 102 | ||
BT132180.1 | - | 2 - 3 | 132 | 102 | ||
BT132273.1 | - | 2 - 3 | 139 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
