Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 13 - 14 | 1087 | 82 | |
XM_015758080.2 | XP_015613566.1 | 13 - 14 | 1025 | 82 | ||
XM_015758081.2 | XP_015613567.1 | 12 - 13 | 925 | 82 | ||
GenBank mRNA | AB039671.1 | - | 13 - 14 | 1065 | 82 | |
AK058373.1 | - | 3 - 4 | 160 | 82 | ||
AK059912.1 | - | 13 - 14 | 883 | 82 | ||
AK068613.1 | - | 13 - 14 | 1005 | 82 | ||
BT128921.1 | - | 3 - 4 | 202 | 82 | ||
BT130454.1 | - | 5 - 6 | 357 | 82 | ||
BT132180.1 | - | 3 - 4 | 187 | 82 | ||
BT132273.1 | - | 3 - 4 | 194 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
