Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 14 - 15 | 1234 | 79 | |
XM_015758080.2 | XP_015613566.1 | 14 - 15 | 1172 | 79 | ||
XM_015758081.2 | XP_015613567.1 | 13 - 14 | 1072 | 79 | ||
GenBank mRNA | AB039671.1 | - | 14 - 15 | 1212 | 79 | |
AK058373.1 | - | 4 - 5 | 307 | 79 | ||
AK059912.1 | - | 14 - 15 | 1030 | 79 | ||
AK068374.1 | - | 14 - 15 | 1227 | 79 | ||
AK068613.1 | - | 14 - 15 | 1152 | 79 | ||
BT128921.1 | - | 4 - 5 | 349 | 79 | ||
BT129662.1 | - | 310 | 79 | |||
BT130454.1 | - | 6 - 7 | 504 | 79 | ||
BT132180.1 | - | 4 - 5 | 334 | 79 | ||
BT132273.1 | - | 4 - 5 | 341 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
