Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 9 - 10 | 808 | 62 | |
XM_015758080.2 | XP_015613566.1 | 9 - 10 | 746 | 62 | ||
XM_015758081.2 | XP_015613567.1 | 8 - 9 | 646 | 62 | ||
GenBank mRNA | AB039671.1 | - | 9 - 10 | 786 | 62 | |
AF251064.1 | - | 7 - 8 | 448 | 62 | ||
AK059912.1 | - | 9 - 10 | 604 | 62 | ||
AK068374.1 | - | 9 - 10 | 801 | 62 | ||
AK068613.1 | - | 9 - 10 | 726 | 62 | ||
BT130454.1 | - | 1 - 2 | 78 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
