Assay Details
| Gene Symbol: LOC4328389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769229.2 | XP_015624715.1 | 2 - 3 | 838 | 83 | |
| XM_015769230.2 | XP_015624716.1 | 3 - 4 | 757 | 83 | ||
| GenBank mRNA | AK105544.1 | - | 3 - 4 | 652 | 83 | |
More Information
Set Membership: |
|

LOC4328389
Genomic Map