Assay Details
Gene Symbol: LOC4328486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770231.2 | XP_015625717.1 | 1 - 2 | 284 | 78 | |
GenBank mRNA | AK119941.1 | - | 1 - 2 | 259 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
