Assay Details
| Gene Symbol: LOC4328524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771577.2 | XP_015627063.1 | 4 - 5 | 823 | 90 | |
| XM_015771578.2 | XP_015627064.1 | 4 - 5 | 515 | 90 | ||
| GenBank mRNA | AK072584.1 | - | 4 - 5 | 712 | 90 | |
| AK121573.1 | - | 3 - 4 | 787 | 90 | ||
More Information
Set Membership: |
|

LOC4328524
Genomic Map