Assay Details
| Gene Symbol: LOC4328524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771577.2 | XP_015627063.1 | 9 - 10 | 1352 | 73 | |
| XM_015771578.2 | XP_015627064.1 | 9 - 10 | 1044 | 73 | ||
| GenBank mRNA | AK072584.1 | - | 9 - 10 | 1241 | 73 | |
| AK121573.1 | - | 8 - 9 | 1311 | 73 | ||
More Information
Set Membership: |
|

LOC4328524
Genomic Map