Assay Details
| Gene Symbol: LOC4329079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768282.2 | XP_015623768.1 | 4 - 5 | 450 | 79 | |
| XM_015768283.2 | XP_015623769.1 | 4 - 5 | 453 | 79 | ||
| XM_015768284.2 | XP_015623770.1 | 3 - 4 | 385 | 79 | ||
| XM_015768285.2 | XP_015623771.1 | 4 - 5 | 452 | 79 | ||
| XM_015768287.2 | XP_015623773.1 | 3 - 4 | 387 | 79 | ||
More Information
Set Membership: |
|

LOC4329079
Genomic Map