Assay Details
Gene Symbol: LOC4329079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768282.2 | XP_015623768.1 | 6 - 7 | 594 | 95 | |
XM_015768283.2 | XP_015623769.1 | 6 - 7 | 597 | 95 | ||
XM_015768284.2 | XP_015623770.1 | 5 - 6 | 529 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
