Assay Details
Gene Symbol: LOC4329079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768282.2 | XP_015623768.1 | 7 - 8 | 700 | 80 | |
XM_015768283.2 | XP_015623769.1 | 7 - 8 | 703 | 80 | ||
XM_015768284.2 | XP_015623770.1 | 6 - 7 | 635 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
