Assay Details
| Gene Symbol: LOC4329079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768282.2 | XP_015623768.1 | 8 - 9 | 887 | 71 | |
| XM_015768284.2 | XP_015623770.1 | 7 - 8 | 822 | 71 | ||
| XM_015768285.2 | XP_015623771.1 | 7 - 8 | 793 | 71 | ||
| XM_015768287.2 | XP_015623773.1 | 6 - 7 | 728 | 71 | ||
More Information
Set Membership: |
|

LOC4329079
Genomic Map