Assay Details
Gene Symbol: LOC4329079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768282.2 | XP_015623768.1 | 9 - 10 | 955 | 86 | |
XM_015768283.2 | XP_015623769.1 | 9 - 10 | 907 | 86 | ||
XM_015768284.2 | XP_015623770.1 | 8 - 9 | 890 | 86 | ||
XM_015768285.2 | XP_015623771.1 | 8 - 9 | 861 | 86 | ||
XM_015768287.2 | XP_015623773.1 | 7 - 8 | 796 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
