Assay Details
Gene Symbol: LOC4329105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768666.2 | XP_015624152.1 | 1 | 760 | 122 | |
GenBank mRNA | AK059847.1 | - | 1 | 751 | 122 | |
AK106082.1 | - | 1 | 752 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
