Assay Details
Gene Symbol: LOC4329669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767689.2 | XP_015623175.1 | 4 - 5 | 933 | 116 | |
XM_015767690.1 | XP_015623176.1 | 4 - 5 | 940 | 116 | ||
GenBank mRNA | AK063555.1 | - | 4 - 5 | 858 | 116 | |
AK067393.1 | - | 4 - 5 | 837 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
