Assay Details
Gene Symbol: LOC4329669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767689.2 | XP_015623175.1 | 6 - 7 | 1349 | 120 | |
XM_015767690.1 | XP_015623176.1 | 6 - 7 | 1356 | 120 | ||
GenBank mRNA | AK063555.1 | - | 6 - 7 | 1274 | 120 | |
AK067393.1 | - | 6 - 7 | 1253 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
