Assay Details
Gene Symbol: LOC4329669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767689.2 | XP_015623175.1 | 7 - 8 | 1737 | 79 | |
GenBank mRNA | AK067393.1 | - | 7 - 8 | 1641 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
