Assay Details
Gene Symbol: LOC4329669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767689.2 | XP_015623175.1 | 9 - 10 | 1989 | 87 | |
XM_015767690.1 | XP_015623176.1 | 9 - 10 | 1990 | 87 | ||
GenBank mRNA | AK067393.1 | - | 9 - 10 | 1893 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
