Assay Details
Gene Symbol: LOC9267717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770923.2 | XP_015626409.1 | 7 - 8 | 771 | 97 | |
XM_015770925.2 | XP_015626411.1 | 6 - 7 | 608 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
