Assay Details
| Gene Symbol: LOC4331551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776716.2 | XP_015632202.1 | 1 - 2 | 158 | 124 | |
| GenBank mRNA | AK060148.1 | - | 1 - 2 | 92 | 124 | |
| AK073584.1 | - | 1 - 2 | 101 | 124 | ||
| AK100231.1 | - | 1 - 2 | 150 | 124 | ||
| AK107864.1 | - | 1 - 2 | 115 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776716.2 transcripts.
Set Membership: |
|

LOC4331551
Genomic Map