Assay Details
| Gene Symbol: LOC4331551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776716.2 | XP_015632202.1 | 4 - 5 | 419 | 110 | |
| XM_015776717.2 | XP_015632203.1 | 4 - 5 | 399 | 110 | ||
| GenBank mRNA | AK060148.1 | - | 4 - 5 | 353 | 110 | |
| AK073584.1 | - | 4 - 5 | 362 | 110 | ||
| AK100231.1 | - | 4 - 5 | 411 | 110 | ||
More Information
Set Membership: |
|

LOC4331551
Genomic Map