Assay Details
| Gene Symbol: LOC4331551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776716.2 | XP_015632202.1 | 5 - 6 | 508 | 80 | |
| XM_015776717.2 | XP_015632203.1 | 5 - 6 | 488 | 80 | ||
| GenBank mRNA | AK060148.1 | - | 5 - 6 | 442 | 80 | |
| AK073584.1 | - | 5 - 6 | 451 | 80 | ||
| AK100231.1 | - | 5 - 6 | 500 | 80 | ||
More Information
Set Membership: |
|

LOC4331551
Genomic Map