Assay Details
Gene Symbol: LOC4331821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777807.2 | XP_015633293.1 | 12 - 13 | 2337 | 101 | |
GenBank mRNA | AK069518.1 | - | 12 - 13 | 2332 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
