Assay Details
Gene Symbol: LOC4331821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777807.2 | XP_015633293.1 | 2 - 3 | 653 | 93 | |
XM_015777808.2 | XP_015633294.1 | 2 - 3 | 653 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
