Assay Details
Gene Symbol: LOC4331821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777807.2 | XP_015633293.1 | 4 - 5 | 986 | 82 | |
XM_015777808.2 | XP_015633294.1 | 4 - 5 | 986 | 82 | ||
GenBank mRNA | AK069518.1 | - | 4 - 5 | 981 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
