Assay Details
Gene Symbol: LOC4331821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777807.2 | XP_015633293.1 | 7 - 8 | 1475 | 79 | |
XM_015777808.2 | XP_015633294.1 | 7 - 8 | 1475 | 79 | ||
GenBank mRNA | AK069518.1 | - | 7 - 8 | 1470 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
