Assay Details
Gene Symbol: LOC4331843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776789.2 | XP_015632275.1 | 2 - 3 | 779 | 145 | |
GenBank mRNA | AK073133.1 | - | 2 - 3 | 719 | 145 | |
AY323478.1 | - | 2 - 3 | 745 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
