Assay Details
Gene Symbol: LOC4332085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776524.2 | XP_015632010.1 | 1 - 2 | 259 | 79 | |
XM_015776525.2 | XP_015632011.1 | 1 - 2 | 259 | 79 | ||
XM_015776526.2 | XP_015632012.1 | 1 - 2 | 259 | 79 | ||
XM_015776527.2 | XP_015632013.1 | 1 - 2 | 259 | 79 | ||
GenBank mRNA | AK059646.1 | - | 1 - 2 | 84 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776524.2, XM_015776525.2, XM_015776526.2, XM_015776527.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
