Assay Details
Gene Symbol: LOC9271214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775313.2 | XP_015630799.1 | 2 - 3 | 641 | 106 | |
XM_015775314.2 | XP_015630800.1 | 2 - 3 | 628 | 106 | ||
GenBank mRNA | AK064767.1 | - | 1 - 2 | 1624 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
