Assay Details
| Gene Symbol: LOC4332728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773854.2 | XP_015629340.1 | 1 - 2 | 245 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332728| Gene Symbol: LOC4332728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773854.2 | XP_015629340.1 | 1 - 2 | 245 | 103 | |
Set Membership: |
|