Assay Details
| Gene Symbol: LOC4332728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773854.2 | XP_015629340.1 | 8 - 9 | 1593 | 80 | |
| XM_015773855.2 | XP_015629341.1 | 7 - 8 | 1290 | 80 | ||
More Information
Set Membership: |
|

LOC4332728
Genomic Map