Assay Details
| Gene Symbol: LOC4332904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777208.2 | XP_015632694.1 | 1 - 2 | 323 | 64 | |
| GenBank mRNA | AK066313.1 | - | 1 - 2 | 321 | 64 | |
| AK066352.1 | - | 1 - 2 | 275 | 64 | ||
More Information
Set Membership: |
|

LOC4332904
Genomic Map