Assay Details
Gene Symbol: LOC4333500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777232.2 | XP_015632718.1 | 14 - 15 | 1700 | 83 | |
XM_015777233.2 | XP_015632719.1 | 14 - 15 | 1705 | 83 | ||
XM_015777234.2 | XP_015632720.1 | 13 - 14 | 1626 | 83 | ||
XM_015777235.2 | XP_015632721.1 | 13 - 14 | 1537 | 83 | ||
XM_015777236.2 | XP_015632722.1 | 13 - 14 | 1631 | 83 | ||
XM_015777238.2 | XP_015632724.1 | 13 - 14 | 1542 | 83 | ||
XM_015777239.2 | XP_015632725.1 | 14 - 15 | 1604 | 83 | ||
GenBank mRNA | AK066665.1 | - | 13 - 14 | 1393 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
