Assay Details
Gene Symbol: LOC4333759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775205.1 | XP_015630691.1 | 3 - 4 | 752 | 73 | |
GenBank mRNA | AK287681.1 | - | 3 - 4 | 758 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
