Assay Details
| Gene Symbol: LOC4333898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777403.2 | XP_015632889.1 | 8 - 9 | 1674 | 115 | |
| GenBank mRNA | BT130148.1 | - | 2 - 3 | 223 | 115 | |
More Information
Set Membership: |
|

LOC4333898
Genomic Map