Assay Details
| Gene Symbol: LOC4334141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775549.2 | XP_015631035.1 | 1 - 2 | 582 | 62 | |
| GenBank mRNA | AB003324.1 | - | 1 - 2 | 441 | 62 | |
| AK100227.1 | - | 1 - 2 | 466 | 62 | ||
| AY551917.1 | - | 1 - 2 | 283 | 62 | ||
| AY551922.1 | - | 1 - 2 | 301 | 62 | ||
| AY551926.1 | - | 1 - 2 | 333 | 62 | ||
More Information
Set Membership: |
|

LOC4334141
Genomic Map