Assay Details
Gene Symbol: LOC4334141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775549.2 | XP_015631035.1 | 4 - 5 | 834 | 129 | |
GenBank mRNA | AB003324.1 | - | 4 - 5 | 693 | 129 | |
AK100227.1 | - | 4 - 5 | 718 | 129 | ||
AY551917.1 | - | 4 - 5 | 535 | 129 | ||
AY551922.1 | - | 4 - 5 | 553 | 129 | ||
AY551926.1 | - | 4 - 5 | 585 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
