Assay Details
| Gene Symbol: LOC4334247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773380.2 | XP_015628866.1 | 1 - 2 | 446 | 119 | |
| GenBank mRNA | AB022675.1 | - | 1 - 2 | 392 | 119 | |
| AK068283.1 | - | 1 - 2 | 448 | 119 | ||
| AK104434.1 | - | 1 - 2 | 441 | 119 | ||
More Information
Set Membership: |
|

LOC4334247
Genomic Map