Assay Details
Gene Symbol: LOC4334611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775269.2 | XP_015630755.1 | 1 - 2 | 1216 | 100 | |
XM_015775270.2 | XP_015630756.1 | 1 - 2 | 1297 | 100 | ||
GenBank mRNA | AK061682.1 | - | 1 - 2 | 304 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
