Assay Details
Gene Symbol: LOC4334611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775269.2 | XP_015630755.1 | 2 - 3 | 1360 | 68 | |
GenBank mRNA | AK061682.1 | - | 2 - 3 | 448 | 68 | |
AK101448.1 | - | 2 - 3 | 452 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
