Assay Details
Gene Symbol: LOC4334611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775269.2 | XP_015630755.1 | 4 - 5 | 1620 | 67 | |
XM_015775270.2 | XP_015630756.1 | 4 - 5 | 1677 | 67 | ||
GenBank mRNA | AK101448.1 | - | 4 - 5 | 712 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
