Assay Details
Gene Symbol: LOC4334611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775269.2 | XP_015630755.1 | 5 - 6 | 1733 | 76 | |
XM_015775270.2 | XP_015630756.1 | 5 - 6 | 1790 | 76 | ||
GenBank mRNA | AK061682.1 | - | 4 - 5 | 599 | 76 | |
AK101448.1 | - | 5 - 6 | 825 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
