Assay Details
Gene Symbol: LOC4335330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777902.2 | XP_015633388.1 | 1 - 2 | 459 | 62 | |
GenBank mRNA | AK069447.1 | - | 1 - 2 | 460 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
