Assay Details
Gene Symbol: LOC4335330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777902.2 | XP_015633388.1 | 4 - 5 | 940 | 106 | |
GenBank mRNA | AK069447.1 | - | 4 - 5 | 941 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
